Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Lipid Quantification Kit, Cell Biolabs

Lipid Quantification Kit, Cell Biolabs

Lipid Quantification Kit, 100 Assays, Fluorometric
Catalog # 102965-236
Marketsource
Supplier:   Cell Biolabs
Supplier #:  STA-617
Lipid Quantification Kit, Cell Biolabs
Catalog # 102965-236
Supplier:  Cell Biolabs
Marketsource
CAS Number:  
Lipid Quantification Kit, Cell Biolabs
Lipid Quantification Kit, Cell Biolabs

Lipid Quantification Kit, 100 Assays, Fluorometric

CAS Number:  
Price
$590.77
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cell Biolabs' Lipid Quantitation Kit provides a simple method for quantitation of unsaturated fatty acids from plasma, serum, or cultured cells in a standard 96-well microplate reader

The kit uses a sulfo-phospho-vanillin system in which samples are acidified and heated to solubilize and prime them for quantitation. In an acidic environment, vanillin reagent reacts with the lipids to form a colored product that is easily detected by the plate reader at 540 nm.

Documents

Specifications

Specifications

  • Description:
    Lipid Quantification Kit
  • Size:
    100 Assays
  • Method:
    Fluorometric
  • Cat. No.:
    102965-236
  • Storage Temperature:
    4°C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.