Lipid Quantification Kit, Cell Biolabs
Lipid Quantification Kit, 100 Assays, Colorimetric
Catalog # 102965-228
Marketsource
Supplier: Cell Biolabs
: STA-613
Lipid Quantification Kit, Cell Biolabs
Catalog # 102965-228
Supplier: Cell Biolabs
Marketsource
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Cell Biolabs' Lipid Quantitation Kit provides a simple method for quantitation of unsaturated fatty acids from plasma, serum, or cultured cells in a standard 96-well microplate reader
The kit uses a sulfo-phospho-vanillin system in which samples are acidified and heated to solubilize and prime them for quantitation. In an acidic environment, vanillin reagent reacts with the lipids to form a colored product that is easily detected by the plate reader at 540 nm.
Specifications
Specifications
- Description:Lipid Quantification Kit
- Size:100 Assays
- Method:Colorimetric
- Cat. No.:102965-228
- Storage Temperature:–20°C



