Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
RIPA Buffer, Cell Biolabs

RIPA Buffer, Cell Biolabs

RIPA Buffer with Protease Inhibitor Cocktail, 20mL
Catalog # 102963-644
Marketsource
Supplier:   Cell Biolabs
Supplier #:  AKR-190
RIPA Buffer, Cell Biolabs
Catalog # 102963-644
Supplier:  Cell Biolabs
Marketsource
CAS Number:  
RIPA Buffer, Cell Biolabs
RIPA Buffer, Cell Biolabs

RIPA Buffer with Protease Inhibitor Cocktail, 20mL

CAS Number:  
Price
$129.23
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cell Biolabs' RIPA Buffer is suitable for the extraction of cytoplasmic, membrane, and nuclear proteins from cells or tissues

The buffer is suitable for a variety of downstream applications such as reporter assays, protein assays, immunoassays and protein purification. RIPA Buffer is available with or without a separate Protease Inhibitor Cocktail.

Documents

Specifications

Specifications

  • Description:
    RIPA Buffer with Protease Inhibitor Cocktail
  • Size:
    20mL
  • Cat. No.:
    102963-644
  • Storage Temperature:
    Room Temperature and –20°C

Specifications