Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
PhosphoBlocker™ Blocking Reagent, Cell Biolabs

PhosphoBlocker™ Blocking Reagent, Cell Biolabs

PhosphoBlocker™ Reagent, 1L
Catalog # 102963-610
Marketsource
Supplier:   Cell Biolabs
Supplier #:  AKR-103
PhosphoBlocker™ Blocking Reagent, Cell Biolabs
Catalog # 102963-610
Supplier:  Cell Biolabs
Marketsource
CAS Number:  
PhosphoBlocker™ Blocking Reagent, Cell Biolabs
PhosphoBlocker™ Blocking Reagent, Cell Biolabs

PhosphoBlocker™ Reagent, 1L

CAS Number:  
Price
$338.46
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

PhosphoBLOCKER™ Blocking Reagent provides superior blocking by maximizing signal-to-noise ratio

The PhosphoBLOCKER™ reagent is an easy-to-use premixed dry blend that enhances low level phosphoprotein signal without increasing background. The reagent particluarly excels with very low levels of endogenous phopsphoproteins.

Most commercially available Western blot blockers, such as dry milk or serum, are sufficient to block unreactive sites on the membrane. However, they are not designed to preserve phosphoprotein antigens during blotting.

Documents

Specifications

Specifications

  • Description:
    PhosphoBlocker™ Reagent
  • Size:
    1L
  • Cat. No.:
    102963-610
  • Storage Temperature:
    RT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.