Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY ACCESSORIES VF2063
Catalog # 10025-182
Supplier:
GREAT LAKES GAGES, LLC
Expand image
VISCOSITY ACCESSORIES VF2063
Catalog # 10025-182
Supplier:
GREAT LAKES GAGES, LLC
Supplier Number:
VF2063