Orders LinkContactUsLinkComponent
Analyzer-Specific Sample Cups, Globe Scientific
Analyzer-Specific Sample Cups, Globe Scientific
Green
Catalog # 10022-136
Supplier:   Globe Scientific
Supplier #:  6500G
Analyzer-Specific Sample Cups, Globe Scientific
Catalog # 10022-136
Supplier:  Globe Scientific
Analyzer-Specific Sample Cups, Globe Scientific
Analyzer-Specific Sample Cups, Globe Scientific
Green
Price
$77.96
Log in for availability
Pack of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Polyethylene sample cups are for use with Cobas Mira® analyzers.
Specifications
  • Color:
    Green