Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STANDARD VISCOSITY N27C SUB-ZERO 1GL
Catalog # 10021-402
Supplier:
Cannon
SDS
Expand image
STANDARD VISCOSITY N27C SUB-ZERO 1GL
Catalog # 10021-402
Supplier:
Cannon
Supplier Number:
9727-G12.128
SDS
Related Information
Documentation
SDS
ghs_9727-G12
Documentation