Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STANDARD VISCOSITY GEN PURPOS N18000 20L
Catalog # 10021-348
Supplier:
Cannon
SDS
Expand image
STANDARD VISCOSITY GEN PURPOS N18000 20L
Catalog # 10021-348
Supplier:
Cannon
Supplier Number:
9727-C63.5
SDS
Related Information
Documentation
SDS
ghs_9727-C63
Documentation