Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STANDARD VISCOSITY GEN PURPOSE S2500 1L
Catalog # 10021-296
Supplier:
Cannon
SDS
Expand image
STANDARD VISCOSITY GEN PURPOSE S2500 1L
Catalog # 10021-296
Supplier:
Cannon
Supplier Number:
9727-C56.032
SDS
Related Information
Documentation
SDS
ghs_9727-C56
Documentation