Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Vitronectin XF™

Vitronectin XF™

Vitronectin XF™, 2 ml
Catalog # MSPP-07180
Marketsource
Supplier #:  07180
Vitronectin XF™
Catalog # MSPP-07180
Marketsource
CAS Number:  
Vitronectin XF™
Vitronectin XF™

Vitronectin XF™, 2 ml

CAS Number:  
Price
$135.92
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Defined, xeno-free matrix that supports the growth and differentiation of human pluripotent stem cells under serum-free, feeder-free conditions.

  • Decrease sources of variability in experiments with a recombinant human protein matrix
  • Handle at room temperature without matrix gelling
  • Use with any TeSR™ family medium to maintain hPSCs
  • Create a completely xeno-free system when used with TeSR™-E8™ or TeSR™-AOF

Support the growth and differentiation of human pluripotent stem cells by using Vitronectin XF™, a defined and xeno-free cell culture matrix.

Developed and manufactured by Nucleus Biologics, Vitronectin XF™ is an effective alternative to Corning® Matrigel®. Use Vitronectin XF™ with mTeSR™1 , mTeSR™ Plus, TeSR™-E8™, or TeSR™-AOF medium to provide a defined culture system for the maintenance of human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells under feeder-free conditions. This system allows complete control over the culture environment, resulting in more consistent cell populations and reproducible results in downstream applications.

Note: CellAdhere™ Dilution Buffer and non-tissue culture-treated cultureware (MSPP-100-0096/MSPP-27147) are required for use with Vitronectin XF™ and are available for purchase separately.

Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Description:
    Vitronectin XF™
  • Size:
    2 ml
  • Cat. No.:
    MSPP-07180
  • Species:
    Human

Specifications