Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Valeric acid ≥99%

Valeric acid ≥99%

100 ml
Catalog # BT135390-100ML
Supplier:   BeanTown Chemical
Supplier #:  135390-100ML
Valeric acid ≥99%
Catalog # BT135390-100ML
Supplier:  BeanTown Chemical
CAS Number:  109-52-4
Valeric acid ≥99%

100 ml

CAS Number:  109-52-4
Price
$26.70
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 109-52-4; EC No: 203-677-2; MDL No: MFCD00004413; RTECS: YV6100000 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Linear Formula: CH3(CH2)3COOH; Molecular Formula: C5H10O2; MW: 102.13 Melting Point: -20 - -18°; Boiling Point: 184-186°; Flash point: 88°C (190°F) Density (g/mL): 0.939; Refractive Index: 1.408

  • RTECS: YV6100000
  • Refractive index: 1.408
  • Liquid
Synonyms:  Pentanoic acid
  • CAS Number:109-52-4
  • Formula:C₅H₁₀O₂
  • MDL Number:MFCD00004413
  • Molecular Weight:102.13 g/mol
  • Boiling Pt:184…186 °C
  • Melting Pt:–20 – –18 °C
  • Density:0.939 g/mL
  • Flash Pt:88 °C (190 °F)
  • EINECS:203-677-2
  • UN:3265
  • ADR:8,III

Documents

Specifications

Specification Test Results

  • Assay:
    99%

Specifications

  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.