1,3-Dioxolane ≥99.5% stabilised
Product Details & Documents
CAS: 646-06-0; EC No: 211-463-5; MDL No: MFCD00003207 UN No: UN1166; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C3H6O2; MW: 74.08 Melting Point: -95°; Boiling Point: 75-76°; Flash point: -3°C (27°F) Density (g/mL): 1.06
- Liquid
- CAS Number:646-06-0
- Formula:C₃H₆O₂
- MDL Number:MFCD00003207
- Molecular Weight:74.08 g/mol
- Boiling Pt:75…76 °C
- Melting Pt:–95 °C
- Density:1.06 g/mL
- Flash Pt:–3 °C (27 °F)
- EINECS:211-463-5
- UN:1166
- ADR:3,II
Specifications
Specification Test Results
- Assay:99.5%
Specifications
- Size:4×1 L
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



