Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Acrylamide ≥99%

Acrylamide ≥99%

500 g
  BT213920-500G
Supplier #:  213920-500G
Acrylamide ≥99%
  BT213920-500G
 :  79-06-1
Acrylamide ≥99%
Acrylamide ≥99%

500 g

 :  79-06-1
 
$126.00
   
Each (500g)
 
   
 

CAS: 79-06-1; EC No: 201-173-7; MDL No: MFCD00008032; RTECS: AS3325000 UN No: UN2074; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: CH2=CHCONH2; Molecular Formula: C3H5NO; MW: 71.08 Melting Point: 82-86°; Boiling Point: 125°/25 mmHg; Flash point: 138°C (280°F) Light Sensitive

  • RTECS: AS3325000
  • Light Sensitive
  • Crystalline
 :  Prop-2-enamide
  •  :79-06-1
  •  :C₃H₅NO
  •  :MFCD00008032
  •  :71.08 g/mol
  •  :125 °C/25 mmHg
  •  :82…86 °C
  •  :138 °C (280 °F)
  •  :201-173-7
  •  :2074
  •  :6.1,III

Documents

 

 

  • Assay:
    99%

 

  • Size:
    500 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.