Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Acrylamide ≥99%

100 g
Catalog # BT213920-100G
Supplier:   BeanTown Chemical
 :  213920-100G
Acrylamide ≥99%
Catalog # BT213920-100G
Supplier:  BeanTown Chemical
CAS Number:  79-06-1
Acrylamide ≥99%
Acrylamide ≥99%

100 g

CAS Number:  79-06-1
Price
$39.00
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 79-06-1; EC No: 201-173-7; MDL No: MFCD00008032; RTECS: AS3325000 UN No: UN2074; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: CH2=CHCONH2; Molecular Formula: C3H5NO; MW: 71.08 Melting Point: 82-86°; Boiling Point: 125°/25 mmHg; Flash point: 138°C (280°F) Light Sensitive

  • RTECS: AS3325000
  • Light Sensitive
  • Crystalline
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Boiling Pt:125 °C/25 mmHg
  • Melting Pt:82…86 °C
  • Flash Pt:138 °C (280 °F)
  • EINECS:201-173-7
  • UN:2074
  • ADR:6.1,III
Specifications

Specification Test Results

  • Assay:
    99%

Specifications

  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.