Acetone ≥99%
Product Details & Documents
CAS: 67-64-1; EC No: 200-662-2; MDL No: MFCD00008765; RTECS: AL3150000 UN No: UN1090; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3COCH3; MW: 58.08 Melting Point: -94°; Boiling Point: 56°; Flash point: -17°C (1°F) Density (g/mL): 0.791; Refractive Index: 1.359
- RTECS: AL3150000
- Refractive index: 1.359
- Liquid
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Boiling Pt:56 °C
- Melting Pt:–94 °C
- Density:0.791 g/mL
- Flash Pt:–17 °C (1 °F)
- EINECS:200-662-2
- UN:1090
- ADR:3,II
Specifications
Specification Test Results
- Assay:99%
Specifications
- Size:2.5 L
- Cat. No.:BT145810-2.5L
- Environmental Certification:Chem21 solvent selection guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



