Ammonia solution 20 - 22%
Product Details & Documents
CAS: 1336-21-6; EC No: 215-647-6; MDL No: MFCD00066650 UN No: UN2672; Haz Class: 8; Packing Group: III Liquid, double sub-boiling quartz distillation; Linear Formula: NH4OH; MW: 35.05 Density (g/mL): 0.9
- Liquid, double sub-boiling quartz distillation
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Density:0.9 g/mL
- EINECS:215-647-6
- UN:2672
- ADR:8,III
Specifications
Specifications
- Size:250 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



