Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

2-Ethoxyethanol ≥99%

2.5 L
  BT133295-2.5L
 :  133295-2.5L
2-Ethoxyethanol ≥99%
  BT133295-2.5L
 :  110-80-5
2-Ethoxyethanol ≥99%
2-Ethoxyethanol ≥99%

2.5 L

 :  110-80-5
 
$90.90
   
Each (2,500ml)
 
   
 

CAS: 110-80-5; EC No: 203-804-1; MDL No: MFCD00002869; RTECS: KK8050000 UN No: UN1171; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3CH2OCH2CH2OH; Molecular Formula: C4H10O2; MW: 90.12 Melting Point: -90°; Boiling Point: 135°; Flash point: 42°C (108°F) Density (g/mL): 0.93; Refractive Index: 1.407

  • RTECS: KK8050000
  • Refractive index: 1.407
  • Liquid
 :  Ethyl glycol, Ethylene glycol monoethyl ether, Cellosolve
  •  :110-80-5
  •  :C₂H₅OCH₂CH₂OH
  •  :MFCD00002869
  •  :90.12 g/mol
  •  :135 °C
  •  :–90 °C
  •  :0.93 g/mL
  •  :42 °C (108 °F)
  •  :203-804-1
  •  :1171
  •  :3,III
 

 

  • Assay:
    99%

 

  • Size:
    2.5 L

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.