Multi-Element QC-21 Check Standard Solution
500 ml, HDPE plastic bottle
Catalog # BT121250-500ML
Supplier: BeanTown Chemical
Supplier #: 121250-500ML
Multi-Element QC-21 Check Standard Solution
Catalog # BT121250-500ML
Supplier: BeanTown Chemical
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:HDPE plastic bottle
- Size:500 ml
- UN No.:3264
- Cat. No.:BT121250-500ML
- MDL No.:MFCD00803610
- GHS signal:Danger
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



