Orders LinkContactUsLinkComponent
Headspace Caps, MicroSolv
Magnetic Crimp Cap, 20 mm, 5 mm Hole, Aluminum, Pharma-fix Grey Butyl Rubber/PTFE Septa, 50 Shore A, 3.0 mm, Gold
Catalog # 10014-050
 :  95025-03-3D
Headspace Caps, MicroSolv
Catalog # 10014-050
Supplier:  
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Magnetic Crimp Cap, 20 mm, 5 mm Hole, Aluminum, Pharma-fix Grey Butyl Rubber/PTFE Septa, 50 Shore A, 3.0 mm, Gold
Discontinued
Product Details & Documents
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Specifications
  • Septum:
    Pharma-fix Grey Butyl Rubber/PTFE Septa, 50 Shore A, 3.0 mm
  • Color:
    Gold
  • Description:
    Magnetic Crimp Cap
  • Material:
    Aluminum
  • Size:
    20 mm, 5 mm Hole