Orders LinkContactUsLinkComponent
dCTP CF® Dye Conjugates, Biotium
dCTP CF® Dye Conjugates, Biotium
CF®488A-dCTP, lyophilized, 490/515 nm, 25 nmol
Catalog # 10001-644
Supplier:   Biotium
Supplier #:  40067
dCTP CF® Dye Conjugates, Biotium
Catalog # 10001-644
Supplier:  Biotium
CAS Number:  
dCTP CF® Dye Conjugates, Biotium
CF®488A-dCTP, lyophilized, 490/515 nm, 25 nmol
CAS Number:  
Price
$500.14
Log in for availability
Each (25mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
CF® dye-dCTP can be used to synthesize probes for in situ hybridization, microarray, or to synthesize labeled DNA probes for in situ hybridization and nucleic acid blotting applications.
  • Synthesize exceptionally bright and photostable fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of nine CF® dye colors from green to far-red
  • Biotiums CF® dyes have superior brightness, photostability, and are water-soluble
Caution: NOTE: CF™594 dCTP is not compatible with PCR labeling of DNA probes.
Specifications
Specifications
  • Description:
    CF®488A-dCTP, lyophilized
  • Size:
    25 nmol
  • Ex/em:
    490/515 nm