PHOSPHATE STD SOLN 30MG/L 946ML
Catalog # 10000-866
Supplier: Hach
Frequently Bought Together
Nalgene® Cryoware Marker Set, Thermo Scientific
Supplier: Thermo Fisher Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Parafilm® M Sealing Film
Supplier: Heathrow Scientific
neodisher® Detergents, Miele
Supplier: Miele
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Sleeve Stoppers
Supplier: VWR International
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Dissecting Scissors, Sharp/Blunt Tip
Supplier: VWR International
N1-METHYLPSEUDOURIDINE-5'-TRIPHOSPHATE
Supplier: TRILINK BIOTECH MS
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Hexane (mixture of isomers) ACS, VWR Chemicals BDH®
Supplier: VWR International
Ethyl acetate ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Dichloromethane ≥99.5% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International