Orders LinkContactUsLinkComponent
Molybdate 3 Reagent Solution, Hach
Molybdate 3 Reagent Solution, Hach
Molybdate 3 Reagent Solution, 1000 mL bottle, Heteropoly blue
Catalog # 10000-792
Molybdate 3 Reagent Solution, Hach
Catalog # 10000-792
Supplier:  
CAS Number:  
Molybdate 3 Reagent Solution, Hach
Molybdate 3 Reagent Solution, Hach
Molybdate 3 Reagent Solution, 1000 mL bottle, Heteropoly blue
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For low and ultra-low range silica determination by the Heteropoly Blue method
Hach methods 8282 and 8186.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Specifications
Specifications
  • Chemical method:
    Heteropoly blue
  • Description:
    Molybdate 3 Reagent Solution
  • Size:
    1000 mL bottle