Orders LinkContactUsLinkComponent
Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit, 11 mm, Yellow, Snap cap
Catalog # 10803-636
Supplier #:  9502C-E-30Y
Caps, BASIK™, MicroSolv
Catalog # 10803-636
Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit, 11 mm, Yellow, Snap cap
Price
$38.99
Log in for availability
Pack of 100
Product Details & Documents
Specifications
  • Cap Size:
    11 mm
  • Cap type:
    Snap cap
  • Septum:
    Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit
  • Material:
    PE
  • Cap color:
    Yellow