Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape), 11 mm, Red, Snap cap
Catalog # 10803-632
Supplier: MicroSolv Technology Corp
Supplier #: 9502C-E-30RX
Jump to:
Product Details & Documents
Specifications
- Cap Size:11 mm
- Cap type:Snap cap
- Septum:Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape)
- Material:PE
- Cap color:Red