Orders LinkContactUsLinkComponent
Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape), 11 mm, Light Blue, Snap cap
Catalog # 10803-626
Supplier #:  9502C-E-30LBXM
Caps, BASIK™, MicroSolv
Catalog # 10803-626
Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape), 11 mm, Light Blue, Snap cap
Price
$414.36
Log in for availability
Case of 1,000
Product Details & Documents
Specifications
  • Cap Size:
    11 mm
  • Cap type:
    Snap cap
  • Septum:
    Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape)
  • Material:
    PE
  • Cap color:
    Light Blue