Orders LinkContactUsLinkComponent
Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit, 11 mm, Black, Snap cap
  10803-614
 :  9502C-E-30K-M
Caps, BASIK™, MicroSolv
  10803-614
Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit, 11 mm, Black, Snap cap
$370.07
Case of 1,000
  • Cap Size:
    11 mm
  • Cap type:
    Snap cap
  • Septum:
    Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit
  • Material:
    PE
  • Cap color:
    Black