Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape), 11 mm, Clear, Snap cap
Catalog # 10803-600
Supplier: MicroSolv Technology Corp
: 9502C-E-30CX
Jump to:
Product Details & Documents
Specifications
- Cap Size:11 mm
- Cap type:Snap cap
- Septum:Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape)
- Material:PE
- Cap color:Clear
- Cat. No.:10803-600