Orders LinkContactUsLinkComponent
Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE, 11 mm, Green, Snap cap
Catalog # 10803-570
Supplier #:  9502C-E-10G-M
Caps, BASIK™, MicroSolv
Catalog # 10803-570
Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE, 11 mm, Green, Snap cap
Price
$335.57
Log in for availability
Case of 1,000
Product Details & Documents
For use with 12 x 32 mm autosampler vials.
Specifications
  • Cap Size:
    11 mm
  • Cap type:
    Snap cap
  • Septum:
    Soft-Guard, White Silicone Rubber / Red PTFE
  • Material:
    PE
  • Cap color:
    Green