Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Adenosine ≥99%

HDPE bottle, 10 g
  BT216680-10G
 :  216680-10G
Adenosine ≥99%
  BT216680-10G
 :  58-61-7
Adenosine ≥99%

HDPE bottle, 10 g

 :  58-61-7
 
$36.70
   
Each (10g)
 
   
 

CAS: 58-61-7; EC No: 200-389-9; MDL No: MFCD00005752; RTECS: AU7175000 Solid; Molecular Formula: C10H13N5O4; MW: 267.24 Melting Point: 234-236°

  • RTECS: AU7175000
  • Solid
 :  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  •  :58-61-7
  •  :C₁₀H₁₃N₅O₄
  •  :MFCD00005752
  •  :267.24 g/mol
  •  :234…236 °C
  •  :200-389-9
 

 

  • Assay:
    99%

 

  • Pack type:
    HDPE bottle
  • Size:
    10 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.