Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Fumaric acid ≥99%

Fumaric acid ≥99%

500 g
Catalog # BT145210-500G
Supplier:   BeanTown Chemical
Supplier #:  145210-500G
Fumaric acid ≥99%
Catalog # BT145210-500G
Supplier:  BeanTown Chemical
CAS Number:  110-17-8
Fumaric acid ≥99%

500 g

CAS Number:  110-17-8
Price
$49.40
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 110-17-8; EC No: 203-743-0; MDL No: MFCD00002700; RTECS: LS9625000 Solid; Linear Formula: HO2CCH=CHCO2H; Molecular Formula: C4H4O4; MW: 116.07 Melting Point: 298-300° (sublimes); Flash point: 230°C (446°F)

  • RTECS: LS9625000
  • Solid
Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116.07 g/mol
  • Melting Pt:298…300 °C (subl.)
  • Flash Pt:230 °C (446 °F)
  • EINECS:203-743-0

Documents

Specifications

Specification Test Results

  • Assay:
    99%

Specifications

  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.