Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Palladium hydroxide ≥99%

Palladium hydroxide ≥99%

5 g
Catalog # BT140370-5G
Supplier:   BeanTown Chemical
Supplier #:  140370-5G
Palladium hydroxide ≥99%
Catalog # BT140370-5G
Supplier:  BeanTown Chemical
CAS Number:  12135-22-7
Palladium hydroxide ≥99%

5 g

CAS Number:  12135-22-7
Price
$808.00
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 12135-22-7; EC No: 235-219-2; MDL No: MFCD00064599 Powder; Linear Formula: Pd(OH)2; MW: 140.42

  • Powder
  • CAS Number:12135-22-7
  • Formula:H₂O₂Pd
  • MDL Number:MFCD00064599
  • Molecular Weight:140.435 g/mol
  • EINECS:235-219-2
Specifications

Specification Test Results

  • Assay:
    99%

Specifications

  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.