Corning® Plain Micro Slides, Corning
Plain slides, 0.9 - 1.1 mm, 75×25 mm (3×1")
Supplier: Corning
: 2947-75X25
Jump to:
Product Details & Documents
For use in nonsterile cytology and pathology applications
Made of water-white glass for maximum clarity. Precleaned and ready for use. Inspected for chips and rough edges before packaging.
: Packaged 1/2 gross per box. Slides are packed vertically for easy lift out.
Certificates
Search by Lot NumberSpecifications
- Description:Plain slides
- L×W:75×25 mm (3×1")
- Thickness:0.9 - 1.1 mm



