Universal Plus Roll, Brady®
Roll, 30W x 150L', 39 gal.
Catalog # 10796-228
Supplier: Brady Worldwide
Supplier #: 107848
Jump to:
Product Details & Documents
Chemically inert making it ideal for use with aggressive fluids
3-ply construction makes it stronger and more durable while reducing lint. The product's green color (the universal symbol for safety) indicates that it is intended for use on hazardous materials. A dimpled roll. Perfed every 12" and up the center to help you use only what you need. For use with chemicals and hazardous fluids.
Documents
Brady GHS Solutions Brochure
Product Literature
Specifications
- Description:Roll
- Dimensions:30W x 150L'
- Absorbency:39 gal.
- Cat. No.:10796-228
- Color:Green
- Class Type:Chemical
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



