Universal Plus Roll, Brady®
Roll, 15W x 150L', 39 gal.
Catalog # 10796-226
Supplier: Brady Worldwide
: 107845
Jump to:
Product Details & Documents
Chemically inert making it ideal for use with aggressive fluids
3-ply construction makes it stronger and more durable while reducing lint. The product's green color (the universal symbol for safety) indicates that it is intended for use on hazardous materials. A dimpled roll. Perfed every 12" and up the center to help you use only what you need. For use with chemicals and hazardous fluids.
Specifications
- Description:Roll
- Dimensions:15W x 150L'
- Absorbency:39 gal.
- Cat. No.:10796-226
- Color:Green
- Class Type:Chemical



