Universal Plus Pad, Brady®
Pad, 12W x 12L", 20 gal.
Catalog # 10796-224
Supplier: Brady Worldwide
Supplier #: 107846
Jump to:
Product Details & Documents
Chemically inert making it ideal for use with aggressive fluids
3-ply construction makes it stronger and more durable while reducing lint. The product's green color (the universal symbol for safety) indicates that it is intended for use on hazardous materials. A dimpled roll. 3-ply, green, dimpled pads. For use with chemicals and hazardous fluids.
Documents
Brady GHS Solutions Brochure
Product Literature
Specifications
- Description:Pad
- Dimensions:12W x 12L"
- Absorbency:20 gal.
- Cat. No.:10796-224
- Color:Green
- Class Type:Chemical



