Universal Plus Pad, Brady®
Pad, 15W x 19L", 20.5 gal.
10796-222
: 107847
Chemically inert making it ideal for use with aggressive fluids
3-ply construction makes it stronger and more durable while reducing lint. The product's green color (the universal symbol for safety) indicates that it is intended for use on hazardous materials. A dimpled roll. 3-ply, green, dimpled pads. For use with chemicals and hazardous fluids.
- Description:Pad
- Dimensions:15W x 19L"
- Absorbency:20.5 gal.
- Cat. No.:10796-222
- Color:Green
- Class Type:Chemical



