Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Can Liners, Elkay Plastics

Star sealed bottom, White, 38×58", 25 µm
Catalog # 10789-604
Supplier:   Elkay Plastics
 :  ALL61WXXX
Can Liners, Elkay Plastics
Catalog # 10789-604
Supplier:  Elkay Plastics
Can Liners, Elkay Plastics
Can Liners, Elkay Plastics

Star sealed bottom, White, 38×58", 25 µm

Price
$112.06
Log in for availability
Case of 100
Product Details & Documents

Can liners have tear and puncture resistance, so they have the strength that is needed to handle the toughest jobs.

They are available in various sizes, thickness and colors.

Specifications
  • Color:
    White
  • Description:
    Star sealed bottom
  • Dimensions:
    38×58"
  • Thickness:
    25 µm