Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Can Liners, Elkay Plastics

Can Liners, Elkay Plastics

Star sealed bottom, White, 15×9×23", 10 µm
Catalog # 10789-560
Supplier:   Elkay Plastics
Supplier #:  ALL24CS
Can Liners, Elkay Plastics
Catalog # 10789-560
Supplier:  Elkay Plastics
Can Liners, Elkay Plastics
Can Liners, Elkay Plastics

Star sealed bottom, White, 15×9×23", 10 µm

Price
$107.88
Log in for availability
Case of 1,000
Product Details & Documents

Can liners have tear and puncture resistance, so they have the strength that is needed to handle the toughest jobs.

They are available in various sizes, thickness and colors.

Specifications
  • Color:
    White
  • Description:
    Star sealed bottom
  • Dimensions:
    15×9×23"
  • Thickness:
    10 µm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.