Orders LinkContactUsLinkComponent
ASTM Class 7 Hooked Weights
0.2 N
Catalog # 10754-376
Supplier:   Troemner
 :  30390783
ASTM Class 7 Hooked Weights
Catalog # 10754-376
Supplier:  Troemner
ASTM Class 7 Hooked Weights
ASTM Class 7 Hooked Weights
0.2 N
Price
$42.16
Log in for availability
Each
Product Details & Documents
ASTM hooked weights for weighing operations in physical and chemical classrooms, such as force measuring.
  • 303 stainless steel
  • ASTM Class 7
All surfaces are satin finish and meet or exceed ANSI/ASTM E617 tolerance and design restrictions.
Specifications
  • Weight:
    0.2 N