Orders LinkContactUsLinkComponent
Corrugated Freezer Boxes
Corrugated Freezer Boxes
Catalog # 10754-062
Corrugated Freezer Boxes
Catalog # 10754-062
Supplier:  
Corrugated Freezer Boxes
Corrugated Freezer Boxes
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FLAT PACK CORUGATED PP FRZR BX NAT PK10 Catalog #10754-060
Product 10754-060 is similar to 10754-062 but manufacturer and specifications may vary.
Product Details & Documents
Easy to assemble corrugated polypropylene boxes are supplied flatpacked to ensure that valuble lab space is not taken up storing boxes before they are needed.
The corrugated polypropylene construction can be frozen and thawed multiple times. Boxes can be stored in temperatures as low as –80 °C (–112°F). The boxes are not susceptible to moisture, which significantly reduces the chance of mold. Storage dividers are included.
Ordering information: Boxes are supplied 10 per pack (10 lids, 10 bases, 10 inserts).