WARMSTART® RTx Reverse Transcriptase, New England Biolabs
WarmStart RTx Reverse Transcriptase, 50, 50 Reactions
Catalog # 102877-822
Supplier: New England Biolabs (NEB)
Supplier #: M0380S
WARMSTART® RTx Reverse Transcriptase, New England Biolabs
Catalog # 102877-822
Supplier: New England Biolabs (NEB)
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
WarmStart RTx Reverse Transcriptase is a unique in silico designed RNA-directed DNA polymerase coupled with a reversibly-bound aptamer that inhibits RTx activity below 40°C
This enzyme can synthesize a complementary DNA strand initiating from a primer using RNA (cDNA synthesis) or single-stranded DNA as a template. RTx is a robust enzyme for RNA detection in amplification reactions and is particularly well suited for use in LAMP (loop-mediated isothermal amplification). The WarmStart property enables high throughput applications, room temperature setup, and increases the consistency and specificity of amplification reactions. RTx contains intact RNase H activity.
Specifications
Specifications
- Description:WarmStart RTx Reverse Transcriptase
- Size:50 Reactions
- No. of reactions:50



