Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Linearity Lipids, AUDIT Microcontrols

Linearity Lipids, AUDIT Microcontrols

Linearity FD lipids for Roche Systems, Freeze dried, Apolipoprotein A-1, Apolipoprotein B, Cholesterol-HDL, Cholesterol-LDL, Cholesterol, Triglycerides, 5 days when stored at 2...8 °C, 5×2 ml
Catalog # 10672-490
Supplier:   Audit Microcontrols
Supplier #:  K882M-5
Linearity Lipids, AUDIT Microcontrols
Catalog # 10672-490
CAS Number:  
Linearity Lipids, AUDIT Microcontrols
Linearity Lipids, AUDIT Microcontrols

Linearity FD lipids for Roche Systems, Freeze dried, Apolipoprotein A-1, Apolipoprotein B, Cholesterol-HDL, Cholesterol-LDL, Cholesterol, Triglycerides, 5 days when stored at 2...8 °C, 5×2 ml

CAS Number:  
Price
$738.40
Log in for availability
Each(2ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Universal linearity lipids, linearity lipids for Roche Systems, and linearity lipids for Beckman AU are assayed quality control materials consisting of five levels of bovine based serum albumin.

Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.

Specifications

Specifications

  • Description:
    Linearity FD lipids for Roche Systems
  • Size:
    5×2 ml
  • Stability:
    5 days when stored at 2...8 °C
  • Analyte:
    Apolipoprotein A-1, Apolipoprotein B, Cholesterol-HDL, Cholesterol-LDL, Cholesterol, Triglycerides
  • Type:
    Freeze dried

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.