Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT HU CXCL11 (I-TAC) (CF) 25UG
Catalog # 10273-572
Supplier:
Tonbo Biosciences
Expand image
RECOMBINANT HU CXCL11 (I-TAC) (CF) 25UG
Catalog # 10273-572
Supplier:
Tonbo Biosciences
Supplier Number:
21-7039-U025