Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Seven2Go™ S8 Pro Portable pH/Ion Meter, METTLER TOLEDO®

Seven2Go™ S8 Pro Portable pH/Ion Meter, METTLER TOLEDO®

Seven2Go™ S8 Pro meter only
Catalog # 10218-844
Supplier:   Mettler Toledo
Supplier #:  30207874
Seven2Go™ S8 Pro Portable pH/Ion Meter, METTLER TOLEDO®
Catalog # 10218-844
Supplier:  Mettler Toledo
Seven2Go™ S8 Pro Portable pH/Ion Meter, METTLER TOLEDO®
Seven2Go™ S8 Pro Portable pH/Ion Meter, METTLER TOLEDO®

Seven2Go™ S8 Pro meter only

Price
$1,110.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

These robust meters provide durability and one-handed operation, suitable for mobile applications in the laboratory, at-line, or outdoors.

  • New intuitive menu guarantees ease of use
  • T-Pad with smart button arrangement enables easy and fast navigation
  • Instrument automatically recognises InLab ISM sensors and stores sensor ID, serial number, and last calibration data
  • Automatic or manual temperature compensation
  • Screen's contrast is enhanced by direct sunlight: Excellent readability even under bright and sunny conditions
  • StatusLight shows the user instantly if the meter is ready or not
  • Data storage of 2000 values (GLP conform)
  • Acoustic endpoint signal

Ordering information: The S8 Pro meter only is supplied with LabX direct pH, USB cable, electrode clip, base accessory, operating instructions, quick guide, test certificate, declaration of conformity and batteries.
S8 Standard Kit: As S8 meter, but also with InLab Expert Pro-ISM-IP 67, bottles for calibration and buffer sachets for pH 4.01, 7.00 and 9.21.
S8 Field Kit:As S8-Standard kit, but also with uGo™ carrying case.
S8 Biotech Kit: As S8-Field kit, but with InLab Routine Pro ISM (incl. cable) instead of InLab Expert Pro-ISM-IP 67.
S8 Fluoride Kit: As S8 Field Kit, but with perfectION Fluoride instead of InLab Expert Pro-ISM-IP 67.

Documents

Specifications
  • Description:
    Seven2Go™ S8 Pro meter only
  • mV range:
    ‒2000.0 to 2000.0
  • Temperature Accuracy:
    ±0.2 °C; ±0.5 °C
  • pH Range:
    –2.00 to 20.00
  • pH Resolution:
    0.001/0.01/0.1
  • Dimensions:
    222W×70D×35H mm (8⁴⁷/₆₄×2³/₄×1³/₈")
  • Power supply:
    4×1.5V Alkaline Batteries or 1.3V Ni-MH Batteries
  • mV resolution:
    0.1/1
  • Weight:
    0.29 kg (0.63 lbs.)
  • pH Accuracy:
    ±0.002
  • Cat. No.:
    10218-844
  • Temperature Compensation:
    Automatic: –5...130 °C (23...266 °F)
    Manual: 30...130 °C (86...266 °F)
  • Outputs:
    Micro USB
  • mV accuracy:
    ±0.1
  • Temperature Resolution:
    0.1 °C
Services

Product Information

  • Catalog #
    Description
    Availability
    Price
  • 10199-716
    IPac standard qualification (IQ/OQ), SevenDirect (installation Pac)
    Log in for availability
    Each - $773.47

  • 10199-718
    EQPac (IQ/OQ/PQ) Comprehensive Qualification, SevenExcellence (Installation Pac)
    Log in for availability
    Each - $1,464.35

Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Benchtop stabilising base, allows users to operate the meter on their lab benches, without the need to hold it in their hands
    Benchtop stabilising base, allows users to operate the meter on their lab benches, without the need to hold it in their hands
    10218-864
    Log in for availability
    Each - $27.80

Product Information

  • Catalog #
    Availability
    Price
  • uGo carrying case, IP67, with sealed interior that allows easy cleaning
    uGo carrying case, IP67, with sealed interior that allows easy cleaning
    10218-862
    Log in for availability
    Each - $249.00

Product Information

  • Catalog #
    Availability
    Price
  • USB cable, 1.2 m, for connection to a PC or to the USB-DC adapter
    USB cable, 1.2 m, for connection to a PC or to the USB-DC adapter
    10218-870
    Log in for availability
    Each - $47.10

Product Information

  • Catalog #
    Availability
    Price
  • DC wallplug adapter for USB cable (includes international converters)
    DC wallplug adapter for USB cable (includes international converters)
    10218-872
    Log in for availability
    Each - $72.60

Product Information

  • Catalog #
    Availability
    Price
  • Wrist strap for meter
    Wrist strap for meter
    10218-868
    Log in for availability
    Each - $23.00

Product Information

  • Catalog #
    Availability
    Price
  • Mini Din-LTW sensor cable adapter
    Mini Din-LTW sensor cable adapter
    11278-286
    Log in for availability
    Each - $162.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.