Tissue-Tek SmartWrite Cassette Printer with Autoloader, Sakura
Thermal transfer printing technology with print that is resistant to histological chemicals
Capable of printing up to 300 dpi to enable high content barcode and alpha numeric characters. Speed of 7.5 seconds per cassette.
Autoloader provides jam-free robotic feed mechanism with 4 clear hoppers for cassettes with or without lids. Each hopper holds 40 cassettes.
Prints on standard smooth-faced cassettes with 35 or 45 degree angle without adjustments.
Output: USB 2.0
: SwartWrite Software (9025), PC with touch screen (9026) and 2-D bar code scanner (9024) are all sold separately.
- Description:Tissue-Tek SmartWrite AutoLoader
- Dimensions:—
- Electrical:100 - 240 VAC, 50/60 Hz
- Includes:Includes one black ribbon, one package of cassettes, 9 cassette hoppers and 9 dividers, printer driver