Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tissue-Tek SmartWrite Cassette Printer with Autoloader, Sakura

Tissue-Tek SmartWrite Cassette Printer with Autoloader, Sakura

Tissue-Tek SmartWrite Cassette Printer (Single), Includes one black ribbon, one package of cassettes, printer driver, 100 - 240 VAC, 50/60 Hz, 22.4 x 31.4 cm
Catalog # 10204-838
Supplier:   Sakura Finetek
Supplier #:  9022
Tissue-Tek SmartWrite Cassette Printer with Autoloader, Sakura
Catalog # 10204-838
Supplier:  Sakura Finetek
Tissue-Tek SmartWrite Cassette Printer with Autoloader, Sakura
Tissue-Tek SmartWrite Cassette Printer with Autoloader, Sakura

Tissue-Tek SmartWrite Cassette Printer (Single), Includes one black ribbon, one package of cassettes, printer driver, 100 - 240 VAC, 50/60 Hz, 22.4 x 31.4 cm

Price
$15,600.00
Log in for availability
Each
Product Details & Documents

Thermal transfer printing technology with print that is resistant to histological chemicals

Capable of printing up to 300 dpi to enable high content barcode and alpha numeric characters. Speed of 7.5 seconds per cassette.

Autoloader provides jam-free robotic feed mechanism with 4 clear hoppers for cassettes with or without lids. Each hopper holds 40 cassettes.

Prints on standard smooth-faced cassettes with 35 or 45 degree angle without adjustments.

Output: USB 2.0

 : SwartWrite Software (9025), PC with touch screen (9026) and 2-D bar code scanner (9024) are all sold separately.

Documents

Specifications
  • Description:
    Tissue-Tek SmartWrite Cassette Printer (Single)
  • Dimensions:
    22.4 x 31.4 cm
  • Electrical:
    100 - 240 VAC, 50/60 Hz
  • Includes:
    Includes one black ribbon, one package of cassettes, printer driver
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • SmartWrite Cassette Magazine
    SmartWrite Cassette Magazine
    10204-842
    Log in for availability
    Case of 4 - $586.00

Product Information

  • Catalog #
    Availability
    Price
  • SmartWrite Cassette Tray Extension (7 to 17 Cassettes)
    SmartWrite Cassette Tray Extension (7 to 17 Cassettes)
    10204-844
    Log in for availability
    Each - $383.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.