Orders LinkContactUsLinkComponent
Staticmaster® EZ Mounts, NRD
Staticmaster® EZ Mounts, NRD
Staticmaster 3" EZ mount for use with 2U500 ionizing cartridge
Catalog # 10183-838
Supplier:   NRD Staticmaster
Supplier #:  BF3
Staticmaster® EZ Mounts, NRD
Catalog # 10183-838
Supplier:  NRD Staticmaster
Staticmaster® EZ Mounts, NRD
Staticmaster® EZ Mounts, NRD
Staticmaster 3" EZ mount for use with 2U500 ionizing cartridge
Price
$70.14
Log in for availability
Each
Product Details & Documents
An indispensable accessory for mounting a 2U500 or 1U400 Ionizing Cartridge in the balance chamber.
  • Simple to use
  • Makes installation of ionizing cartridges fast and easy
Simply remove the protective layer of the double-sided tape and affix the E-Z mount to the back wall of the chamber. Once the mount is attached, installing ionizing cartridges is fast and easy.
Specifications
  • Description:
    Staticmaster 3" EZ mount for use with 2U500 ionizing cartridge