Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Trypsin EDTA Solution, PromoCell

Trypsin EDTA Solution, PromoCell

Trypsin/EDTA (0.04%/0.03%), 250 ml
Catalog # 10172-552
Supplier:   Promocell
Supplier #:  C-41020
Trypsin EDTA Solution, PromoCell
Catalog # 10172-552
Supplier:  Promocell
CAS Number:  
Trypsin EDTA Solution, PromoCell

Trypsin/EDTA (0.04%/0.03%), 250 ml

CAS Number:  
Price
$94.82
Log in for availability
Each (250ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Trypsin is the most widely employed enzyme used for cell detachment.

  • Available in different sizes
  • Store at –20 °C in the dark
  • Thawed solutions can be stored at 4 to 8 °C

PromoCell's Trypsin Solutions exhibit standardized enzyme activity and contain trypsin from extra-pure lots.

All lots have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Caution: The products are for in vitro research use only and not for diagnostic or therapeutic procedures.

Specifications

Specifications

  • Description:
    Trypsin/EDTA (0.04%/0.03%)
  • Volume:
    250 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.