Orders LinkContactUsLinkComponent
Indicator Strips
Indicator Strips
Indicator strips, Dry heat
Catalog # 10163-884
Supplier:   Sklar
Supplier #:  10-1117
Indicator Strips
Catalog # 10163-884
Supplier:  Sklar
Indicator Strips
Indicator Strips
Indicator strips, Dry heat
Price
$80.97
Log in for availability
Box of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Disposable paper indicator strips are latex-free and nonsterile.
Specifications
  • Description:
    Indicator strips
  • Legend:
    Dry heat