Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Glass Jugs, Amber

Glass Jugs, Amber

3.8 L (130 oz.), 38-400, Black Phenolic with Pulp/Vinyl Liner, —
Catalog # 16142-402
Supplier:   Qorpak
Supplier #:  GLC-02054
Glass Jugs, Amber
Catalog # 16142-402
Supplier:  Qorpak
Glass Jugs, Amber
Glass Jugs, Amber

3.8 L (130 oz.), 38-400, Black Phenolic with Pulp/Vinyl Liner, —

Price
$217.76
Log in for availability
Case of 6
Product Details & Documents

Amber glass jugs with molded finger grips are the perfect storage solution for light sensitive products.

Amber glass protects against harmful UV rays.

Documents

Specifications
  • Cap Size:
    38-400
  • Cap type:
    Black Phenolic with Pulp/Vinyl Liner
  • Capacity:
    3.8 L (130 oz.)
  • Treatment:
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.